KEY414334571 Key Organics / BIONET

1-{[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}piperidine

  • MDL number MFCD08435871
  • Cas Number : 859833-21-9
  • Molecular Weight : 301.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/402927

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2757_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart