KEY414334575 Key Organics / BIONET

benzyl 4-{[(4-methylbenzene)sulfonyl]oxy}piperidine-1-carboxylate

  • MDL number MFCD00065304
  • Cas Number : 196601-12-4
  • Molecular Weight : 389.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/409260

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-3431_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart