KEY414334583 Key Organics / BIONET

1,1-diethylpropargylamine

  • MDL number MFCD00008057
  • Cas Number : 3234-64-8
  • Molecular Weight : 111.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1,1-diethyl-2-propynylamine
Inchi Code 1S/C7H13N/c1-4-7(8,5-2)6-3/h1H,5-6,8H2,2-3H3
Inchi Key WHNOKDNCUNZBLW-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Liquid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/409267

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-4705_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart