KEY414334589 Key Organics / BIONET

ethyl 2-(2-methyl-4-oxo-4,5,6,7-tetrahydro-1H-indol-1-yl)acetate

  • MDL number MFCD15474013
  • Cas Number : 1260673-29-7
  • Molecular Weight : 235.283

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/409273

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4711_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart