KEY414334592 Key Organics / BIONET

ethyl 1,2-dimethyl-4-oxo-4,5,6,7-tetrahydro-1H-1,3-benzodiazole-6-carboxylate

  • MDL number MFCD27924240
  • Cas Number : 871724-23-1
  • Molecular Weight : 236.271

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/409276

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4714_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart