KEY414334596 Key Organics / BIONET

methyl 3-(benzyloxy)-5-hydroxybenzoate

  • MDL number MFCD06655985
  • Cas Number : 54915-31-0
  • Molecular Weight : 258.273

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/409280

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4720_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart