KEY414334598 Key Organics / BIONET

N-(3,5-difluorophenyl)-2,2,2-trifluoroacetamide

  • MDL number MFCD20546924
  • Cas Number : 1378573-08-0
  • Molecular Weight : 225.118

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/409282

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4722_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart