KEY414334599 Key Organics / BIONET

(1S,2R,6R,8R,9R)-9-hydroxy-4,4-dimethyl-3,5,7,11- tetraoxatricyclo[6.3.0.0^{2,6}]undecan-10-one

  • MDL number MFCD08461814
  • Cas Number : 29514-28-1
  • Molecular Weight : 216.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/409283

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-4723_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart