KEY414334606 Key Organics / BIONET

5-chloro-1-[(4-methylbenzene)sulfonyl]-3-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine

  • MDL number MFCD16039195
  • Cas Number : 866546-11-4
  • Molecular Weight : 432.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/409290

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4730_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart