KEY415137536 Key Organics / BIONET

(9H-fluoren-9-yl)methyl N-[(2R)-1-oxo-3-[(triphenylmethyl)sulfanyl]propan-2-yl]carbamate

  • MDL number MFCD27957305
  • Cas Number : 505076-77-7
  • Molecular Weight : 569.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 58 - 62
Purity 95
Storage temp. Store under N2 at -18°C
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/427464

Safety Information

signalword Warning
pcodes P411,P422, P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AK-0714_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart