KEY415137539 Key Organics / BIONET

tetrabutylazanium; trifluoro({3-[(4-methylpiperazin-1-yl)methyl]phenyl})boranuide

  • MDL number MFCD27957317
  • Molecular Weight : 499.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/427517

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2846_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart