KEY415137543 Key Organics / BIONET

2-(5-bromo-2-methoxyphenyl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione

  • MDL number MFCD27957323
  • Cas Number : 1287221-38-8
  • Molecular Weight : 341.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/427537

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2866_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart