KEY415137544 Key Organics / BIONET

(1S,2R,7S)-9-methyl-3-oxa-9-azatricyclo[3.3.1.0^{2,4}]nonan-7-yl (2S)-3-hydroxy-2-phenylpropanoate trihydrate hydrobromide

  • Cas Number : 114-49-8
  • Molecular Weight : 438.315

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/427538

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2867_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart