KEY415137551 Key Organics / BIONET

2-bromo-N-(2,3-dihydro-1H-inden-5-yl)-2-phenylacetamide

  • MDL number MFCD27980585
  • Cas Number : 864932-47-8
  • Molecular Weight : 330.225

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 116 - 118
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/427450

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11T-1540_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart