KEY415137554 Key Organics / BIONET

methoxy(4-methoxyphenyl)dimethylsilane

  • MDL number MFCD09266147
  • Cas Number : 62244-48-8
  • Molecular Weight : 196.321

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/427467

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2172_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart