KEY415137555 Key Organics / BIONET

sodium dimethyl(thiophen-2-yl)silanolate

  • MDL number MFCD09266152
  • Cas Number : 879904-87-7
  • Molecular Weight : 180.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store under N2
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/427468

Safety Information

signalword Warning
pcodes P422, P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2190_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart