KEY415137557 Key Organics / BIONET

2-{4-[(2-ethylhexyl)oxy]phenyl}-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD22415440
  • Cas Number : 251566-01-5
  • Molecular Weight : 332.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/427470

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2497_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart