KEY415137567 Key Organics / BIONET

ethyl 4-(3-amino-4-nitrophenyl)piperazine-1-carboxylate

  • MDL number MFCD27957307
  • Cas Number : 23470-45-3
  • Molecular Weight : 294.311

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/427480

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2769_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart