KEY415137568 Key Organics / BIONET

N1-[2-(dimethylamino)ethyl]-4-nitrobenzene-1,3-diamine

  • MDL number MFCD27957308
  • Cas Number : 1339047-06-1
  • Molecular Weight : 224.264

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/427481

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2770_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart