KEY415137569 Key Organics / BIONET

2-[4-(3-amino-4-nitrophenyl)piperazin-1-yl]ethan-1-ol

  • MDL number MFCD00545981
  • Cas Number : 23470-44-2
  • Molecular Weight : 266.301

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/427482

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2771_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart