KEY415137572 Key Organics / BIONET

methyl 4-{[4-(trifluoromethyl)pyridin-2-yl]oxy}benzoate

  • MDL number MFCD27957294
  • Cas Number : 869109-13-7
  • Molecular Weight : 297.233

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 76 - 78
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/427451

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11Z-0868_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart