KEY415137578 Key Organics / BIONET

4-hydroxy-4'-methyl-[1,1'-biphenyl]-3-carboxylic acid

  • MDL number MFCD00434467
  • Cas Number : 43029-70-5
  • Molecular Weight : 228.247

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/427491

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2780_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart