KEY415137589 Key Organics / BIONET

methyl 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalene-2-carboxylate

  • MDL number MFCD18783177
  • Cas Number : 736989-93-8
  • Molecular Weight : 312.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-naphthoate
Inchi Code 1S/C18H21BO4/c1-17(2)18(3,4)23-19(22-17)15-9-8-12-10-14(16(20)21-5)7-6-13(12)11-15/h6-11H,1-5H3
Inchi Key WEDOOEMXFXWHAU-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/427502

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-2792_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart