KEY415137595 Key Organics / BIONET

tert-butyl 4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzenesulfonyl]piperazine-1-carboxylate

  • MDL number MFCD14702533
  • Cas Number : 1042917-53-2
  • Molecular Weight : 452.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/427508

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2798_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart