KEY415137603 Key Organics / BIONET

2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

  • MDL number MFCD16996321
  • Cas Number : 870238-67-8
  • Molecular Weight : 247.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/427516

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2844_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart