KEY415137604 Key Organics / BIONET

methyl 2-(pyridin-2-yl)-4-(pyridin-4-yl)pyrimidine-5-carboxylate

  • MDL number MFCD27957296
  • Cas Number : 1565845-69-3
  • Molecular Weight : 292.298

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156 - 158
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/427453

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1E-903_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart