KEY415137615 Key Organics / BIONET

potassium trifluoro[4-(2-methoxy-2-oxoethoxy)phenyl]boranuide

  • MDL number MFCD27957320
  • Molecular Weight : 272.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/427528

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2857_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart