KEY415137624 Key Organics / BIONET

methyl 4-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]methoxy}benzoate

  • MDL number MFCD27957299
  • Cas Number : 1565827-80-6
  • Molecular Weight : 345.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147 - 149
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/427457

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3P-099_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart