KEY415137625 Key Organics / BIONET

ethyl [(3-carbamoylthiophen-2-yl)carbamoyl]formate

  • MDL number MFCD03012541
  • Cas Number : 68746-57-6
  • Molecular Weight : 242.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 185
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73230

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3T-0832_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart