KEY415137657 Key Organics / BIONET

ethyl 6-chloro-5-methylpyridine-3-carboxylate

  • MDL number MFCD20485595
  • Cas Number : 942511-70-8
  • Molecular Weight : 199.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp LMS
Iupac Name ethyl 6-chloro-5-methylnicotinate
Inchi Code 1S/C9H10ClNO2/c1-3-13-9(12)7-4-6(2)8(10)11-5-7/h4-5H,3H2,1-2H3
Inchi Key ARMQAPYXINXSLX-UHFFFAOYSA-N

Description

Melting Point LMS
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/427570

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/MH-0728_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart