KEY415137667 Key Organics / BIONET

methyl 5-(4-chloro-3-methylphenyl)-1H-pyrazole-3-carboxylate

  • MDL number MFCD26387491
  • Cas Number : 192702-05-9
  • Molecular Weight : 250.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/431078

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4739_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart