KEY415137670 Key Organics / BIONET

5,5-dimethyl-3',3'a,4',6'a-tetrahydro-1'H-spiro[1,3-dioxane-2,2'-pentalene]

  • MDL number MFCD27957338
  • Cas Number : 92007-41-5
  • Molecular Weight : 208.301

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/431081

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4742_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart