KEY415137673 Key Organics / BIONET

(1S)-2,2-dimethyl-1-phenylpropane-1,3-diol

  • MDL number MFCD24444156
  • Cas Number : 133164-39-3
  • Molecular Weight : 180.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/431084

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-4745_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart