KEY415137683 Key Organics / BIONET

(2S)-3-[(4-methoxyphenyl)methoxy]-2-methylpropan-1-ol

  • MDL number MFCD27957342
  • Cas Number : 160238-45-9
  • Molecular Weight : 210.273

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/431094

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4755_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart