KEY415137686 Key Organics / BIONET

5-[(2-chlorophenyl)carbonyl]-1,3-thiazol-2-amine

  • MDL number MFCD26393360
  • Cas Number : 848462-68-0
  • Molecular Weight : 238.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/431097

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4758_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart