KEY415137688 Key Organics / BIONET

{1-[(4-fluorophenyl)methyl]-1H-pyrazol-4-yl}methanol

  • MDL number MFCD16862192
  • Cas Number : 1094202-70-6
  • Molecular Weight : 206.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/431099

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4760_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart