KEY415137691 Key Organics / BIONET

2,4-dibromonaphthalen-1-amine

  • MDL number MFCD00155345
  • Cas Number : 20191-76-8
  • Molecular Weight : 300.981

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/431102

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4764_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart