KEY415137693 Key Organics / BIONET

5,5-dimethyl-hexahydro-1'H-spiro[1,3-dioxane-2,2'-pentalene]-5'-one

  • MDL number MFCD27957346
  • Cas Number : 92007-37-9
  • Molecular Weight : 224.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/431104

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4766_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart