KEY423214580 Key Organics / BIONET

2-methyl-1,3-benzoxazole-6-carboxylic acid

  • MDL number MFCD07787266
  • Cas Number : 13452-14-7
  • Molecular Weight : 177.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-methyl-1,3-benzoxazole-6-carboxylic acid
Inchi Code 1S/C9H7NO3/c1-5-10-7-3-2-6(9(11)12)4-8(7)13-5/h2-4H,1H3,(H,11,12)
Inchi Key GFYDDTFAVDJJAI-UHFFFAOYSA-N

Description

Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/431134

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/4W-0404_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart