KEY423214600 Key Organics / BIONET

2-methyl-5-nitro-1,2-dihydroisoquinolin-1-one

  • MDL number MFCD00489134
  • Cas Number : 42792-96-1
  • Molecular Weight : 204.185

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114 - 116
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/431125

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1C-003_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart