KEY423214604 Key Organics / BIONET

2-oxo-2-phenylethyl 2-[(4-chlorophenyl)carbamoyl]acetate

  • MDL number MFCD27980586
  • Cas Number : 1582770-07-7
  • Molecular Weight : 331.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139 - 141
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/431126

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1G-056_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart