KEY423214610 Key Organics / BIONET

5-(6-methyl-4,8-dioxo-1,3,6,2-dioxazaborocan-2-yl)thiophene-2-carbaldehyde

  • MDL number MFCD27980601
  • Cas Number : 1104637-06-0
  • Molecular Weight : 267.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/431173

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2891_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart