KEY423214611 Key Organics / BIONET

3-amino-1-(4-nitrophenyl)urea

  • MDL number MFCD00041266
  • Cas Number : 17433-93-1
  • Molecular Weight : 196.166

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/431127

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1X-0324_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart