KEY423214618 Key Organics / BIONET

1-(2-{[3-(trifluoromethyl)phenyl]amino}-1,3-thiazol-5-yl)ethan-1-one

  • MDL number MFCD27980587
  • Cas Number : 1582770-06-6
  • Molecular Weight : 286.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 193 - 195
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/431128

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2T-0885_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart