KEY423214634 Key Organics / BIONET

methyl 2-hydroxynaphthalene-1-carboxylate

  • MDL number MFCD00454146
  • Cas Number : 947-65-9
  • Molecular Weight : 202.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 79-81�c
bp 330.5�c at 760mmHg
Iupac Name methyl 2-hydroxy-1-naphthoate
Inchi Code 1S/C12H10O3/c1-15-12(14)11-9-5-3-2-4-8(9)6-7-10(11)13/h2-7,13H,1H3
Inchi Key LEENMPKUUNPLHM-UHFFFAOYSA-N

Description

Melting Point 79-81�c
Purity 97%
Physical Form Solid
Country of Origin GB
Boiling point 330.5�c at 760mmHg
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/431197

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-7015_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart