KEY423214640 Key Organics / BIONET

3-(morpholin-4-yl)propanoic acid hydrobromide

  • MDL number MFCD03452744
  • Cas Number : 28025-77-6
  • Molecular Weight : 240.097

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/431203

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-7031_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart