KEY423214671 Key Organics / BIONET

methyl (2S)-2-amino-3-(5-hydroxy-1H-indol-3-yl)propanoate hydrochloride

  • MDL number MFCD00036995
  • Cas Number : 60971-91-7
  • Molecular Weight : 270.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150 decomp
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/431239

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CK-0703_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart