KEY423214698 Key Organics / BIONET

2-butyl-1-benzofuran-5-carboxylic acid

  • MDL number MFCD22575243
  • Cas Number : 1447931-31-8
  • Molecular Weight : 218.252

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/431265

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/RS-1009_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart