KEY423214703 Key Organics / BIONET

[1-(4-chlorophenyl)cyclopropyl]methanol

  • MDL number MFCD00066929
  • Cas Number : 80866-81-5
  • Molecular Weight : 182.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/431270

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/RS-1014_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart