KEY423214704 Key Organics / BIONET

4-cyanobenzamide

  • MDL number MFCD00017133
  • Cas Number : 3034-34-2
  • Molecular Weight : 146.149

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/431271

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/RS-1015_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart